Learn More
Medchemexpress LLC Beta-glucuronidase (helix pomatia) | 9001-45-0 | 500 KU
SDP
Catalog No. 5000399901
Encompass_Preferred
Beta-glucuronidase (helix pomatia) is a glycosyl hydrolase that functions by hydrolyzing β-glucuronic acid and sulfate esters. This process occurs in biological fluids such as urine, leading to the release of β-glucuronate. It is intended for research use only and is not sold to patients. The product is available in solid form, appearing white to off-white in color, and is supplied with a data sheet, SDS, COA, and handling instructions.
  • Enzymatic function: It is a glycosyl hydrolase.
  • Substrates: Hydrolyzes β-glucuronic acid and sulfate esters.
  • Resulting product: Releases β-glucuronate.
  • Applications: Used in research.
  • Physical appearance: Solid, white to off-white.
Catalog No. 50-003-99901 Supplier Medchemexpress LLC Supplier No. HYP2803B500KU
Beta-glucuronidase (helix pomatia) is a glycosyl hydrolase that functions by hydrolyzing β-glucuronic acid and sulfate esters. This process occurs in biological fluids such as urine, leading to the release of β-glucuronate. It is intended for research use only and is not sold to patients. The product is available in solid form, appearing white to off-white in color, and is supplied with a data sheet, SDS, COA, and handling instructions.
  • Enzymatic function: It is a glycosyl hydrolase.
  • Substrates: Hydrolyzes β-glucuronic acid and sulfate esters.
  • Resulting product: Releases β-glucuronate.
  • Applications: Used in research.
  • Physical appearance: Solid, white to off-white.