Learn More
TARGETMOL CHEMICALS INC BETA-GLUCURONIDASE-IN-1 5MG
SDP
Catalog No. 502581342
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. (beta)-Glucuronidase-IN-1 is an E. coli (beta)-glucuronidase ((beta)-glucuronidase) inhibitor with a potent selective non-competitive and orally active IC50 and Ki value of 283 nM and 164 nM respectively against E. coli (beta)-glucuronidase. purity: 98%
Catalog No. 50-258-1342 Supplier TARGETMOL CHEMICALS INC Supplier No. T134775MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. (beta)-Glucuronidase-IN-1 is an E. coli (beta)-glucuronidase ((beta)-glucuronidase) inhibitor with a potent selective non-competitive and orally active IC50 and Ki value of 283 nM and 164 nM respectively against E. coli (beta)-glucuronidase. purity: 98%