Learn More
Cell Biolabs Inc Beta-Hexosaminidase Assay Kit, 100 assays
Catalog No. NC1634727
Encompass_Preferred
Hexosaminidase is an enzyme that hydrolyzes terminal N-acetyl-D-hexosamine residues from N-acetyl-ß-D-hexosaminidines. The enzyme is typically found as a dimer of the alpha and beta subunits. Cell Biolabs' Beta Hexosaminidase Assay Kit quantifies the activity of the beta subunit in a convenient 96-well microplate format. Each kit provides sufficient reagents for 100 assays including standards and unknown samples.
Catalog No. NC1634727 Supplier Cell Biolabs Inc Supplier No. MET5095
Hexosaminidase is an enzyme that hydrolyzes terminal N-acetyl-D-hexosamine residues from N-acetyl-ß-D-hexosaminidines. The enzyme is typically found as a dimer of the alpha and beta subunits. Cell Biolabs' Beta Hexosaminidase Assay Kit quantifies the activity of the beta subunit in a convenient 96-well microplate format. Each kit provides sufficient reagents for 100 assays including standards and unknown samples.