Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vidan™ Diagnostics Beta-hydroxybutyrate (BHB) Linearity Set

Catalog No. 23300113
Encompass_Preferred
Change view
Click to view available options
Quantity:
1
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-300-113 1
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-300-113 Supplier Vidan™ Diagnostics Supplier No. 137209943052
This item is not returnable. View return policy

Beta-hydroxybutyrate (BHB) Linearity Set to be used for validation of linear range of the Beta-hydroxybutyrate 21FS Reagent Kit

  • Linearity Set for Beta-hydroxybuyrate (BHB), (6 x 4ml)
  • Keep refrigerated
  • 60 day open vial stability
  • Perform Linearity Check as recommended by your institution
  • FDA Cleared

Specifications

Certifications/Compliance FDA cleared
Content And Storage Refrigerated
Concentration 6 levels of BHB standards
Detectable Analytes beta-hydroxybutyrate (BHB)
For Use With (Equipment) Open System Clinical Chemistry Analyzers
Type BHB Linearity Standards
Includes 6 x 4 mL (levels 1 to 6)
Level 1 to 6
Quantity 1
Source Water Based
Stability until expiration if properly stored
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.