Learn More
DIASYS DIAGNOSTIC SYSTEMS USA LLC DiaSys β-Hydroxybutyrate FS is a ready-to-use two component enzymatic test to determine β-Hydroxybutyrate in serum or heparin plasma on photometric systems. Each set contains R1= 90mL, R2=24mL
Catalog No. NC2874345
Encompass_Preferred
DiaSys β-Hydroxybutyrate 21 FS is an enzymatic test to determine β-hydroxybutyrate in serum or plasma on photometric systems. For veterinary or research use only. Increased β-hydroxybutyrate levels indicate metabolic acidosis related to diabetes, congenital metabolic diseases, alcoholism or fasting. The reagent is liquid-stable with long onboard and calibration stabilities. Quantitative results ensure early intervention. Measurement of β-Hydroxybutyrate is superior to urine ketone testing. Method: Enzymatic determination with β-Hydroxybutyrate-Dehydrogenase.Reagent 1 contains Buffer, and β=Hydroxybutyrate-Dehydrogenase, reagent 2 contains Buffer, and NAD. Reagents must be stored refrigerated.
Catalog No. NC2874345 Supplier DIASYS DIAGNOSTIC SYSTEMS USA LLC Supplier No. 137019911921
DiaSys β-Hydroxybutyrate 21 FS is an enzymatic test to determine β-hydroxybutyrate in serum or plasma on photometric systems. For veterinary or research use only. Increased β-hydroxybutyrate levels indicate metabolic acidosis related to diabetes, congenital metabolic diseases, alcoholism or fasting. The reagent is liquid-stable with long onboard and calibration stabilities. Quantitative results ensure early intervention. Measurement of β-Hydroxybutyrate is superior to urine ketone testing. Method: Enzymatic determination with β-Hydroxybutyrate-Dehydrogenase.Reagent 1 contains Buffer, and β=Hydroxybutyrate-Dehydrogenase, reagent 2 contains Buffer, and NAD. Reagents must be stored refrigerated.