Learn More
DIASYS DIAGNOSTIC SYSTEMS USA LLC DiaSys β-Hydroxybutyrate is an aqueous Standard for use with β-Hydroxybutyrate asays (3x3mL)
Catalog No. NC2874344
Encompass_Preferred
DiaSys β-Hydroxybutyrate Standard is an aqueous standard for use in tests for quantitative in vitro determination of β-Hydroxybutyrate. For veterinary or research use only. The standard is used to calibrate β-Hydroxybutyrate assays. DiaSys β-Hydroxybutyrate Standard, both opened and unopened, must be stored refrigerated. Avoid contamination and protect from light. Once opened, DiaSys β-Hydroxybutyrate Standard is stable for 24 months if kept refrigerated and recapped immediatley after use.
Catalog No. NC2874344 Supplier DIASYS DIAGNOSTIC SYSTEMS USA LLC Supplier No. 137009911030
DiaSys β-Hydroxybutyrate Standard is an aqueous standard for use in tests for quantitative in vitro determination of β-Hydroxybutyrate. For veterinary or research use only. The standard is used to calibrate β-Hydroxybutyrate assays. DiaSys β-Hydroxybutyrate Standard, both opened and unopened, must be stored refrigerated. Avoid contamination and protect from light. Once opened, DiaSys β-Hydroxybutyrate Standard is stable for 24 months if kept refrigerated and recapped immediatley after use.