Learn More
Medchemexpress LLC Beta-hydroxyisovalerylshikonin | 7415-78-3 | 1 MG
SDP
Catalog No. 5000419274
Encompass_Preferred
Beta-hydroxyisovalerylshikonin is a natural product isolated from *Lithospermum erythrorhizon*. It acts as a potent inhibitor of protein tyrosine kinases (PTK) and is effective against a wide variety of tumor cell lines.
  • Potent inhibitor of protein tyrosine kinases (PTK)
  • Effective against various tumor cell lines
  • Induces cell death in NCI-H522 and DMS114 cells
  • For research use only
Catalog No. 50-004-19274 Supplier Medchemexpress LLC Supplier No. HYN42011MG
Add to Cart
Add to Cart
Beta-hydroxyisovalerylshikonin is a natural product isolated from *Lithospermum erythrorhizon*. It acts as a potent inhibitor of protein tyrosine kinases (PTK) and is effective against a wide variety of tumor cell lines.
  • Potent inhibitor of protein tyrosine kinases (PTK)
  • Effective against various tumor cell lines
  • Induces cell death in NCI-H522 and DMS114 cells
  • For research use only