Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Beta III Tubulin Antibody | 50 kDa | 10 UL
SDP

Catalog No. 5000397584
Encompass_Preferred

Beta III Tubulin Antibody is a non-conjugated Rabbit IgG monoclonal antibody that targets beta III Tubulin. It can be used as a loading control antibody and is suitable for various applications.

  • Reactivity: human, mouse, rat
  • Host: rabbit
  • Clonality: monoclonal
  • Conjugation: non-conjugated
  • Molecular weight: 50 kDa (predicted band size)
  • Formulation: supplied in 50 mM Tris-Glycine (pH 7.4), 0.15 M NaCl, 40% Glycerol and 0.05% BSA
  • Preservative: 0.01% Sodium azide

Catalog No. 50-003-97584 Supplier Medchemexpress LLC Supplier No. HYP8057010UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Beta III Tubulin Antibody is a non-conjugated Rabbit IgG monoclonal antibody that targets beta III Tubulin. It can be used as a loading control antibody and is suitable for various applications.

  • Reactivity: human, mouse, rat
  • Host: rabbit
  • Clonality: monoclonal
  • Conjugation: non-conjugated
  • Molecular weight: 50 kDa (predicted band size)
  • Formulation: supplied in 50 mM Tris-Glycine (pH 7.4), 0.15 M NaCl, 40% Glycerol and 0.05% BSA
  • Preservative: 0.01% Sodium azide

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.