Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC beta III Tubulin Antibody (YA4243) | 50 kDa | 50 UL
SDP

Catalog No. 5000446200
Encompass_Preferred

The beta III Tubulin Antibody (YA4243) is a mouse-derived, non-conjugated IgG1 monoclonal antibody from MedChemExpress. It targets TUBB3 and is provided as a liquid at a concentration of 1 mg/mL. This antibody is suitable for various research applications such as Western Blot, Immunocytochemistry/Immunofluorescence, Flow Cytometry, and ELISA.

  • Host: Mouse
  • Reactivity: Human, Mouse, Rat
  • Clonality: YA4243 monoclonal antibody
  • Molecular weight: 50 kDa
  • Appearance: Liquid
  • Concentration: 1 mg/mL
  • Conjugation: Non-conjugated
  • Applications: WB, ICC/IF, FC, ELISA
  • Formulation: Supplied in PBS with 0.05% sodium azide
  • Storage: Store at -20°C for 1 year; avoid repeated freeze/thaw cycles
  • Shipping: With blue ice
  • For research use only

Catalog No. 50-004-46200 Supplier Medchemexpress LLC Supplier No. HYP8454650UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The beta III Tubulin Antibody (YA4243) is a mouse-derived, non-conjugated IgG1 monoclonal antibody from MedChemExpress. It targets TUBB3 and is provided as a liquid at a concentration of 1 mg/mL. This antibody is suitable for various research applications such as Western Blot, Immunocytochemistry/Immunofluorescence, Flow Cytometry, and ELISA.

  • Host: Mouse
  • Reactivity: Human, Mouse, Rat
  • Clonality: YA4243 monoclonal antibody
  • Molecular weight: 50 kDa
  • Appearance: Liquid
  • Concentration: 1 mg/mL
  • Conjugation: Non-conjugated
  • Applications: WB, ICC/IF, FC, ELISA
  • Formulation: Supplied in PBS with 0.05% sodium azide
  • Storage: Store at -20°C for 1 year; avoid repeated freeze/thaw cycles
  • Shipping: With blue ice
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.