Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Beta III Tubulin Antibody (YA4243) | Predicted band size: 50 kDa; Observed band size: 50 kDa | 100 UL
SDP

Catalog No. 5000446199
Encompass_Preferred

This beta III Tubulin Antibody (YA4243) is supplied as a liquid solution with a concentration of 1 mg/mL. It has a predicted and observed molecular weight of 50 kDa and is stable for 1 year when stored at -20°C. It is designed for laboratory research applications.

  • Host: Mouse
  • Reactivity: Human, Mouse, Rat
  • Conjugation: Non-conjugated
  • Applications: WB, ICC/IF, FC, ELISA
  • Formulation: Supplied in PBS with 0.05% sodium azide
  • Storage: Stored at -20°C for 1 year; avoid repeated freeze/thaw cycles

Catalog No. 50-004-46199 Supplier Medchemexpress LLC Supplier No. HYP84546100UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This beta III Tubulin Antibody (YA4243) is supplied as a liquid solution with a concentration of 1 mg/mL. It has a predicted and observed molecular weight of 50 kDa and is stable for 1 year when stored at -20°C. It is designed for laboratory research applications.

  • Host: Mouse
  • Reactivity: Human, Mouse, Rat
  • Conjugation: Non-conjugated
  • Applications: WB, ICC/IF, FC, ELISA
  • Formulation: Supplied in PBS with 0.05% sodium azide
  • Storage: Stored at -20°C for 1 year; avoid repeated freeze/thaw cycles

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.