Learn More
Medchemexpress LLC Beta III Tubulin Antibody (YA586) | 50 kDa | 100 UL
SDP
Catalog No. 5000396404
Encompass_Preferred
This product is a non-conjugated monoclonal antibody, originating from rabbits, that targets beta III Tubulin. It can be utilized as a loading control antibody.
  • Specifically targets beta III Tubulin.
  • Acts as a reliable loading control antibody.
  • Provides broad species reactivity including human, mouse, and rat.
Catalog No. 50-003-96404 Supplier Medchemexpress LLC Supplier No. HYP80570100UL
This product is a non-conjugated monoclonal antibody, originating from rabbits, that targets beta III Tubulin. It can be utilized as a loading control antibody.
  • Specifically targets beta III Tubulin.
  • Acts as a reliable loading control antibody.
  • Provides broad species reactivity including human, mouse, and rat.