Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Beta III Tubulin Antibody (YA586) | 50 kDa | 50 UL
SDP

Catalog No. 5000396449
Encompass_Preferred

This beta III Tubulin Antibody is a non-conjugated, rabbit-originated IgG monoclonal antibody targeting beta III Tubulin. It functions as a loading control antibody. Beta III Tubulin is a class III member of the beta tubulin protein family, primarily expressed in neurons, and plays a role in neurogenesis, axon guidance, and maintenance.

  • Host: Rabbit
  • Reactivity: Human, Mouse, Rat
  • Concentration: 0.3 mg/mL
  • Conjugation: Non-conjugated
  • Molecular weight: 50 kDa
  • Purity: Affinity purified
  • Storage: Stored at -20°C for 1 year
  • Appearance: Liquid
  • Applications: WB, IHC-F, IHC-P, ICC/IF, IP

Catalog No. 50-003-96449 Supplier Medchemexpress LLC Supplier No. HYP8057050UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This beta III Tubulin Antibody is a non-conjugated, rabbit-originated IgG monoclonal antibody targeting beta III Tubulin. It functions as a loading control antibody. Beta III Tubulin is a class III member of the beta tubulin protein family, primarily expressed in neurons, and plays a role in neurogenesis, axon guidance, and maintenance.

  • Host: Rabbit
  • Reactivity: Human, Mouse, Rat
  • Concentration: 0.3 mg/mL
  • Conjugation: Non-conjugated
  • Molecular weight: 50 kDa
  • Purity: Affinity purified
  • Storage: Stored at -20°C for 1 year
  • Appearance: Liquid
  • Applications: WB, IHC-F, IHC-P, ICC/IF, IP

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.