Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Beta-ionone | 14901-07-6 | MFCD00001549 | 99.9% | 192.30 g·mol⁻¹ | C13H20O | 200 MG
SDP

Catalog No. 5000302613
Encompass_Preferred

β-Ionone is a small aromatic ketone (C13H20O) used as a research reagent and analytical standard. It is a light yellow to colorless liquid and has reported activity inducing apoptosis in gastric adenocarcinoma SGC7901 cells.

  • CAS number: 14901-07-6
  • Molecular formula: C13H20O
  • Molecular weight: 192.30 g·mol⁻¹
  • Purity: ~99.9% (high purity for research)
  • Available pack sizes: 10 mM (1 mL in DMSO), 100 mg, 200 mg, 500 mg, 1 g
  • Typical applications: apoptosis research and analytical assays

Catalog No. 50-003-02613 Supplier Medchemexpress LLC Supplier No. HYW015084200MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

β-Ionone is a small aromatic ketone (C13H20O) used as a research reagent and analytical standard. It is a light yellow to colorless liquid and has reported activity inducing apoptosis in gastric adenocarcinoma SGC7901 cells.

  • CAS number: 14901-07-6
  • Molecular formula: C13H20O
  • Molecular weight: 192.30 g·mol⁻¹
  • Purity: ~99.9% (high purity for research)
  • Available pack sizes: 10 mM (1 mL in DMSO), 100 mg, 200 mg, 500 mg, 1 g
  • Typical applications: apoptosis research and analytical assays

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.