Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Beta-ionone | 14901-07-6 | MFCD00078759 | 99.9% | 192.30 g/mol | C13H20O | 100 MG
SDP

Catalog No. 5000303949
Encompass_Preferred

β-Ionone is a small organic compound (C13H20O, MW 192.30 g/mol) used in research as an apoptosis inducer with reported anti-cancer activity in gastric adenocarcinoma SGC7901 cells. It is supplied as solid and solution formats for in vitro and in vivo studies and includes solubility and storage recommendations for reliable handling.

  • High purity (≈99.85%).
  • Available as solid and DMSO solution formats for experimental use.
  • Soluble in DMSO and ethanol; insoluble in water.
  • Storage recommendations for pure form and for solutions to preserve stability.
  • Acts as an apoptosis inducer in cell studies.
  • Molecular formula C13H20O; molecular weight 192.30 g/mol.

Catalog No. 50-003-03949 Supplier Medchemexpress LLC Supplier No. HYW015084100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

β-Ionone is a small organic compound (C13H20O, MW 192.30 g/mol) used in research as an apoptosis inducer with reported anti-cancer activity in gastric adenocarcinoma SGC7901 cells. It is supplied as solid and solution formats for in vitro and in vivo studies and includes solubility and storage recommendations for reliable handling.

  • High purity (≈99.85%).
  • Available as solid and DMSO solution formats for experimental use.
  • Soluble in DMSO and ethanol; insoluble in water.
  • Storage recommendations for pure form and for solutions to preserve stability.
  • Acts as an apoptosis inducer in cell studies.
  • Molecular formula C13H20O; molecular weight 192.30 g/mol.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.