Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Beta-ionone | 79-77-6 | MFCD00001549 | 96.8% | 192.30 g/mol | C13H20O | 1 ML
SDP

Catalog No. 5000302497
Encompass_Preferred

(E)-β-Ionone is the trans isomer of β-ionone, an apocarotenoid with reported pro-apoptotic activity in cellular studies. Supplied as a ready-to-use 10 mM solution in DMSO or as powdered material, it is intended for research applications.

  • E-isomer of β-ionone with reported pro-apoptotic activity in cell studies.
  • Available as a 10 mM solution in DMSO and as solid packs (25-500 mg).
  • High purity, approximately 96.8%.
  • Chemical formula C13H20O; molecular weight 192.30 g/mol.
  • Good solubility in DMSO; in vivo formulation protocols provided.
  • Storage stability: room temperature for solid; long-term storage in solvent at -80°C.

Catalog No. 50-003-02497 Supplier Medchemexpress LLC Supplier No. HYW015084A10MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(E)-β-Ionone is the trans isomer of β-ionone, an apocarotenoid with reported pro-apoptotic activity in cellular studies. Supplied as a ready-to-use 10 mM solution in DMSO or as powdered material, it is intended for research applications.

  • E-isomer of β-ionone with reported pro-apoptotic activity in cell studies.
  • Available as a 10 mM solution in DMSO and as solid packs (25-500 mg).
  • High purity, approximately 96.8%.
  • Chemical formula C13H20O; molecular weight 192.30 g/mol.
  • Good solubility in DMSO; in vivo formulation protocols provided.
  • Storage stability: room temperature for solid; long-term storage in solvent at -80°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.