Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Beta IV Tubulin Antibody (YA2267) | 10 UL
SDP

Catalog No. 5000443094
Encompass_Preferred

beta IV Tubulin Antibody (YA2267) is a rabbit-derived non-conjugated IgG monoclonal antibody that specifically targets beta IV Tubulin. This antibody can be utilized as a loading control and is reactive with human, mouse, and rat samples. It is supplied as a liquid formulation with a predicted and observed molecular weight of 50 kDa.

  • Rabbit-derived non-conjugated IgG monoclonal antibody
  • Targets beta IV Tubulin
  • Can be used as a loading control antibody
  • Host: Rabbit
  • Reactivity: Human, mouse, rat
  • Conjugation: Non-conjugated
  • SwissProt ID: P04350
  • Research field: Tags & cell markers
  • Appearance: Liquid

Catalog No. 50-004-43094 Supplier Medchemexpress LLC Supplier No. HYP8252210UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

beta IV Tubulin Antibody (YA2267) is a rabbit-derived non-conjugated IgG monoclonal antibody that specifically targets beta IV Tubulin. This antibody can be utilized as a loading control and is reactive with human, mouse, and rat samples. It is supplied as a liquid formulation with a predicted and observed molecular weight of 50 kDa.

  • Rabbit-derived non-conjugated IgG monoclonal antibody
  • Targets beta IV Tubulin
  • Can be used as a loading control antibody
  • Host: Rabbit
  • Reactivity: Human, mouse, rat
  • Conjugation: Non-conjugated
  • SwissProt ID: P04350
  • Research field: Tags & cell markers
  • Appearance: Liquid

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.