Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Beta IV Tubulin Antibody (YA2267) | 50 UL
SDP

Catalog No. 5000444470
Encompass_Preferred

Beta IV Tubulin Antibody (YA2267) is a rabbit-derived, non-conjugated IgG monoclonal antibody that targets beta IV Tubulin. This antibody can be used as a loading control. Beta IV Tubulin is a primary component of microtubules, which are cylindrical structures formed by alpha- and beta-tubulin heterodimers. Microtubules elongate by adding GTP-tubulin dimers to their ends, creating a stabilizing cap.

  • Host: Rabbit
  • Isotype: IgG monoclonal antibody
  • Reactivity with human, mouse, and rat samples
  • Suitable for Western Blot (WB), Immunohistochemistry-Paraffin (IHC-P), Immunohistochemistry-Frozen (IHC-F), and Immunocytochemistry/Immunofluorescence (ICC/IF) applications
  • Molecular weight: 50 kDa (predicted and observed)
  • Storage conditions: -20°C for 1 year, avoid repeated freeze/thaw cycles

Catalog No. 50-004-44470 Supplier Medchemexpress LLC Supplier No. HYP8252250UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Beta IV Tubulin Antibody (YA2267) is a rabbit-derived, non-conjugated IgG monoclonal antibody that targets beta IV Tubulin. This antibody can be used as a loading control. Beta IV Tubulin is a primary component of microtubules, which are cylindrical structures formed by alpha- and beta-tubulin heterodimers. Microtubules elongate by adding GTP-tubulin dimers to their ends, creating a stabilizing cap.

  • Host: Rabbit
  • Isotype: IgG monoclonal antibody
  • Reactivity with human, mouse, and rat samples
  • Suitable for Western Blot (WB), Immunohistochemistry-Paraffin (IHC-P), Immunohistochemistry-Frozen (IHC-F), and Immunocytochemistry/Immunofluorescence (ICC/IF) applications
  • Molecular weight: 50 kDa (predicted and observed)
  • Storage conditions: -20°C for 1 year, avoid repeated freeze/thaw cycles

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.