Learn More
Medchemexpress LLC Beta-lactamase CTX-M 20ug
SDP
Catalog No. 5000196084
Encompass_Preferred
Recombinant CTX-M-1 beta-lactamase expressed in E. coli with an N-terminal His tag. Supplied as a purified protein for research applications including enzyme characterization, inhibitor screening, and assay development. The product is provided in a 20 μg pack, with purity >90% (reducing SDS-PAGE) and an approximate molecular weight of 32.2 kDa; store frozen and aliquot after reconstitution.
  • High purity (>90%) as determined by reducing SDS-PAGE.
  • E. coli expression system suitable for bacterial protein studies.
  • N-terminal His tag enables affinity purification and detection.
  • Stability when stored frozen; aliquot for long-term storage at -80°C.
  • For research use only; not for human therapeutic or diagnostic use.
Catalog No. 50-001-96084 Supplier Medchemexpress LLC Supplier No. HYP7145920UG
Recombinant CTX-M-1 beta-lactamase expressed in E. coli with an N-terminal His tag. Supplied as a purified protein for research applications including enzyme characterization, inhibitor screening, and assay development. The product is provided in a 20 μg pack, with purity >90% (reducing SDS-PAGE) and an approximate molecular weight of 32.2 kDa; store frozen and aliquot after reconstitution.
  • High purity (>90%) as determined by reducing SDS-PAGE.
  • E. coli expression system suitable for bacterial protein studies.
  • N-terminal His tag enables affinity purification and detection.
  • Stability when stored frozen; aliquot for long-term storage at -80°C.
  • For research use only; not for human therapeutic or diagnostic use.