Learn More
Medchemexpress LLC Beta-lactamase CTX-M 50ug
SDP
Catalog No. 5000195675
Encompass_Preferred
Recombinant Beta-lactamase CTX-M-1 (His tag) is a purified, E. coli-expressed enzyme that hydrolyzes penicillins and cephalosporins and is supplied as a small-scale research reagent for biochemical assays and inhibitor testing.
  • Recombinant CTX-M-1 enzyme with N-terminal His tag.
  • Expressed in Escherichia coli for consistent production.
  • Supplied as a 50 μg purified preparation with >90% purity.
  • Suitable for enzymatic assays and inhibitor screening.
  • Supports studies of extended-spectrum β-lactamase activity and resistance mechanisms.
Catalog No. 50-001-95675 Supplier Medchemexpress LLC Supplier No. HYP7145950UG
Recombinant Beta-lactamase CTX-M-1 (His tag) is a purified, E. coli-expressed enzyme that hydrolyzes penicillins and cephalosporins and is supplied as a small-scale research reagent for biochemical assays and inhibitor testing.
  • Recombinant CTX-M-1 enzyme with N-terminal His tag.
  • Expressed in Escherichia coli for consistent production.
  • Supplied as a 50 μg purified preparation with >90% purity.
  • Suitable for enzymatic assays and inhibitor screening.
  • Supports studies of extended-spectrum β-lactamase activity and resistance mechanisms.