Learn More
Sigma Aldrich Fine Chemicals Biosciences beta-Lactoglobulin from bo1G
Catalog No. 502897968
Encompass
Milk from dairy cows contains the protein -lactoglobulin (BLG). It naturally occurs in a number of genetic variants and the most prevalent bovine variants are BLG A and BLG B.
Catalog No. 50-289-7968 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. L01301G
Milk from dairy cows contains the protein -lactoglobulin (BLG). It naturally occurs in a number of genetic variants and the most prevalent bovine variants are BLG A and BLG B.