Learn More
Sigma Aldrich Fine Chemicals Biosciences beta Lactoglobulin from bo5G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences L39085G
Encompass
Milk from dairy cows contains the protein -lactoglobulin (BLG). It naturally occurs in a number of genetic variants and the most prevalent bovine variants are BLG A and BLG B.
Catalog No. 50-290-7340