Learn More
Selleck Chemical LLC Beta-Lapachone 10mg 4707-32-8 -Lapachone, ARQ-501
SDP
Supplier:  Selleck Chemical LLC S726110MG
Encompass_Preferred
Beta-Lapachone (?-Lapachone, ARQ-501) is a selective DNA topoisomerase I inhibitor, exhibiting no inhibitory activities against DNA topoisomerase II or ligase. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4569