Learn More
TARGETMOL CHEMICALS INC BETA-LAPACHONE 50MG
SDP
Catalog No. 502579385
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. (beta)-Lapachone (ARQ-501) is a specific DNA topoisomerase I inhibitor and no inhibitory activities against DNA topoisomerase II or ligase. purity: 99%
Catalog No. 50-257-9385 Supplier TARGETMOL CHEMICALS INC Supplier No. T640750MG
Also available in 5 mg 10 mg 25 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. (beta)-Lapachone (ARQ-501) is a specific DNA topoisomerase I inhibitor and no inhibitory activities against DNA topoisomerase II or ligase. purity: 99%