Learn More
Enzo Life Sciences β-Lapachone (5mg). CAS: 4707-32-8
SDP
Supplier:  Enzo Life Sciences BMLGR3080005
Encompass_Preferred
Novel DNA topoisomerase I inhibitor. Unlike camptothecin, β-lapachone does not stabilize the cleavable complex, indicating a novel mode of action. Induces apoptosis in HL-60 and human prostate cancer cells via a p53-independent mechanism and blocks the cell cycle at G0/G1. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO (25mg/ml) or ethanol (20mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-0958