Learn More
Selleck Chemical LLC Beta-Lapachone
SDP
Supplier:  Selleck Chemical LLC S726150MG
Encompass_Preferred
Beta-Lapachone ( -Lapachone ARQ-501) is a selective DNA topoisomerase I inhibitor exhibiting no inhibitory activities against DNA topoisomerase II or ligase Phase 2
Catalog No. 50-000-00831