Learn More
Selleck Chemical LLC Beta-Lapachone
SDP
Catalog No. 5000004521
Encompass_Preferred
Beta-Lapachone ( -Lapachone ARQ-501) is a selective DNA topoisomerase I inhibitor exhibiting no inhibitory activities against DNA topoisomerase II or ligase Phase 2
Catalog No. 50-000-04521 Supplier Selleck Chemical LLC Supplier No. S726110MM/1ML
Beta-Lapachone ( -Lapachone ARQ-501) is a selective DNA topoisomerase I inhibitor exhibiting no inhibitory activities against DNA topoisomerase II or ligase Phase 2