Learn More
Abcam beta-Lapachone, Topoisomerase I inhibitor, 10MG
Catalog No. 502706906
Encompass_Preferred
MW 244.28 Da, Purity >98%. Topoisomerase I inhibitor. Inhibits topoisomerase I induced DNA cleavage. Induces apoptosis via a p53 independent mechanism. Shows antitumor, antiviral, antibacterial, antifungal and wound healing activities.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6906 Supplier Abcam Supplier No. AB14109710MG
MW 244.28 Da, Purity >98%. Topoisomerase I inhibitor. Inhibits topoisomerase I induced DNA cleavage. Induces apoptosis via a p53 independent mechanism. Shows antitumor, antiviral, antibacterial, antifungal and wound healing activities.
The product is subject to the following: Abcam Restricted Use Statement