Learn More
Abcam beta-Lapachone, Topoisomerase I inhibitor, 50MG
Catalog No. 502706905
Encompass_Preferred
MW 244.28 Da, Purity >98%. Topoisomerase I inhibitor. Inhibits topoisomerase I induced DNA cleavage. Induces apoptosis via a p53 independent mechanism. Shows antitumor, antiviral, antibacterial, antifungal and wound healing activities.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6905 Supplier Abcam Supplier No. AB14109750MG
MW 244.28 Da, Purity >98%. Topoisomerase I inhibitor. Inhibits topoisomerase I induced DNA cleavage. Induces apoptosis via a p53 independent mechanism. Shows antitumor, antiviral, antibacterial, antifungal and wound healing activities.
The product is subject to the following: Abcam Restricted Use Statement