Learn More
Apexbio Technology LLC Beta-Lipotropin (1-10), porcine 77875-68-4 25mg
SDP
Catalog No. 5000093133
Encompass_Preferred
Beta-Lipotropin 1-10 porcine is an endogenous opioid peptide fragment derived from the N-terminal region of beta-lipotropin It is structurally related to opioid peptides and interacts with opioid receptors to mediate morphine-like activity within the central nervous system Beta-Lipotropin 1-10 porcine exerts its biological effects through modulation of opioid receptor signaling pathways Based on these pharmacological properties Beta-Lipotropin 1-10 porcine holds research potential in neuropharmacological studies particularly in investigations related to endogenous pain modulation neurotransmission mechanisms and opioid receptor ligand interactions
Catalog No. 50-000-93133 Supplier Apexbio Technology LLC Supplier No. A101425
Beta-Lipotropin 1-10 porcine is an endogenous opioid peptide fragment derived from the N-terminal region of beta-lipotropin It is structurally related to opioid peptides and interacts with opioid receptors to mediate morphine-like activity within the central nervous system Beta-Lipotropin 1-10 porcine exerts its biological effects through modulation of opioid receptor signaling pathways Based on these pharmacological properties Beta-Lipotropin 1-10 porcine holds research potential in neuropharmacological studies particularly in investigations related to endogenous pain modulation neurotransmission mechanisms and opioid receptor ligand interactions