Learn More
Medchemexpress LLC Beta-Mangostin | 20931-37-7 | 424.49 | 5 MG
SDP
Catalog No. 5000415587
Encompass_Preferred
beta-Mangostin is a xanthone compound found in *Cratoxylum arborescens*, known for its antibacterial and antimalarial properties. It exhibits antimycobacterial activity against *Mycobacterium tuberculosis* and *in vitro* antimalarial activity against *Plasmodium falciparum*. Additionally, beta-Mangostin demonstrates potent anticancer activity against various cancers, including hepatocellular carcinoma and leukaemia, by activating the intrinsic apoptosis pathway and inducing G0/G1 cell-cycle arrest.
  • Exhibits antibacterial activity against *Mycobacterium tuberculosis*
  • Shows antimalarial activity against *Plasmodium falciparum*
  • Possesses potent anticancer activity against various cancers
  • Activates the intrinsic apoptosis pathway
  • Induces G0/G1 cell-cycle arrest in HL60 cells
Catalog No. 50-004-15587 Supplier Medchemexpress LLC Supplier No. HYN09415MG
beta-Mangostin is a xanthone compound found in *Cratoxylum arborescens*, known for its antibacterial and antimalarial properties. It exhibits antimycobacterial activity against *Mycobacterium tuberculosis* and *in vitro* antimalarial activity against *Plasmodium falciparum*. Additionally, beta-Mangostin demonstrates potent anticancer activity against various cancers, including hepatocellular carcinoma and leukaemia, by activating the intrinsic apoptosis pathway and inducing G0/G1 cell-cycle arrest.
  • Exhibits antibacterial activity against *Mycobacterium tuberculosis*
  • Shows antimalarial activity against *Plasmodium falciparum*
  • Possesses potent anticancer activity against various cancers
  • Activates the intrinsic apoptosis pathway
  • Induces G0/G1 cell-cycle arrest in HL60 cells