Learn More
Medchemexpress LLC beta-Mangostin | 20931-37-7 | MFCD00951751 | 424.49 | 1 MG
SDP
Catalog No. 5000415345
Encompass_Preferred
beta-Mangostin is a xanthone compound found in *Cratoxylum arborescens* with antibacterial and antimalarial activities. It also demonstrates potent anticancer activity against various cancers, activating the intrinsic apoptosis pathway and causing G0/G1 cell-cycle arrest.
  • Exhibits antimycobacterial activity against *Mycobacterium tuberculosis*.
  • Shows antimalarial activity against *Plasmodium falciparum*.
  • Possesses anticancer activity against hepatocellular carcinoma and leukemic cells.
  • Activates intrinsic apoptosis pathway.
  • Associated with G0/G1 cell-cycle arrest.
Catalog No. 50-004-15345 Supplier Medchemexpress LLC Supplier No. HYN09411MG
beta-Mangostin is a xanthone compound found in *Cratoxylum arborescens* with antibacterial and antimalarial activities. It also demonstrates potent anticancer activity against various cancers, activating the intrinsic apoptosis pathway and causing G0/G1 cell-cycle arrest.
  • Exhibits antimycobacterial activity against *Mycobacterium tuberculosis*.
  • Shows antimalarial activity against *Plasmodium falciparum*.
  • Possesses anticancer activity against hepatocellular carcinoma and leukemic cells.
  • Activates intrinsic apoptosis pathway.
  • Associated with G0/G1 cell-cycle arrest.