Learn More
TARGETMOL CHEMICALS INC Beta-mangostin 5MG
SDP
Catalog No. 502880214
Encompass_Preferred
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Beta-mangostin (beta-Mangostin) is a xanthone compound with antibacterial and antimalarial activities. Beta-mangostin present in Cratoxylum arborescens and exhibits antimycobacterial activity against Mycobacterium tuberculosis with an MIC of 6.25 ug/mL. Beta-mangostin possesses in vitro antimalarial activity against Plasmodium falciparum, with an IC50 of 3.00 ug/mL. Beta-mangostin has potent anticancer activity against various cancers (such as hepatocellular carcinoma, leukaemic). Purity 99.69%
Catalog No. 50-288-0214 Supplier TARGETMOL CHEMICALS INC Supplier No. TN10195MG
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Beta-mangostin (beta-Mangostin) is a xanthone compound with antibacterial and antimalarial activities. Beta-mangostin present in Cratoxylum arborescens and exhibits antimycobacterial activity against Mycobacterium tuberculosis with an MIC of 6.25 ug/mL. Beta-mangostin possesses in vitro antimalarial activity against Plasmodium falciparum, with an IC50 of 3.00 ug/mL. Beta-mangostin has potent anticancer activity against various cancers (such as hepatocellular carcinoma, leukaemic). Purity 99.69%