Learn More
AdipoGen beta-Mannosylceramide (250 µg)
SDP
Supplier:  AdipoGen AGCR13621C250
Encompass_Preferred
Chemical. CAS: 1334332-65-8. Formula: C50H99NO9. MW: 858.3. Potent stimulator of invariant natural killer T (iNKT) cells. Induces strong antitumor immunity through a nitric oxide (NO) and TNF-alpha dependent mechansim, different compared to the IFN-gamma dependent mechanism of alpha-GalCer (AG-CN2-0014). Does not induce anergy of iNKT cells.
Catalog No. 50-205-3990