Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beta Max™-Esliquid Scintillation Cocktail, MP Biomedicals™

Catalog No. MP188002005 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 gal.
4 x 1 gal.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP188002005 5 gal.
MP188002004 4 x 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP188002005 Supplier MP Biomedicals, Inc Supplier No. 0188002005
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Features high tritium efficiency and is excellent with dry filters and test wipes. MP Biomedicals Beta Max™-Esliquid Scintillation Cocktail is available in a large, 5 gallon option or in four 1 gallon containers.

  • Biodegradable, non-toxic, non-flammable and odorless
  • Product can be applied with organic samples, dry filters, test wipes and 3H Acetyl CoA CAT assay extractions
  • Store at room temperature (15–30°C)
TRUSTED_SUSTAINABILITY

Specifications

Boiling Point 300°C
Density 0.9 - 1.0g/mL at 20°C
Quantity 5 gal.
Recommended Storage Store at Room Temperature (15 - 30°C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.