Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals beta-Methyl-DL-phenylalanine hydrochloride, 99%

Catalog No. AC202660010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC202660010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC202660010 Supplier Thermo Scientific Chemicals Supplier No. 202660010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 80997-87-1 | C10H14ClNO2 | 215.68 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80997-87-1
Molecular Formula C10H14ClNO2
Molecular Weight (g/mol) 215.68
MDL Number MFCD00012623
InChI Key SGKWQBMDOXGYAA-UHFFFAOYNA-N
Synonym 2-amino-3-phenylbutanoic acid hydrochloride, apba, 2-amino-3-phenylbutanoic acid, chloride, beta-methyl-3-phenyl-dl-alanine hcl, beta-methyl-3-phenyl-dl-alanine hydrochloride, acmc-20ao59, beta-methyl dl phenylalanine hydrochloride, beta-methyl-dl-phenylalanine hydrochloride, 2-azanyl-3-phenyl-butanoic acid hydrochloride, phenylalanine, b-methyl-, hydrochloride 1:1
PubChem CID 12743546
IUPAC Name 2-amino-3-phenylbutanoic acid;hydrochloride
SMILES Cl.CC(C(N)C(O)=O)C1=CC=CC=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Linear Formula C6H5CH(CH3)CH(NH2)CO2H·HCl
Quantity 1 g
Formula Weight 215.68
Percent Purity ≥98.5%
Physical Form Powder
Chemical Name or Material β-Methyl-DL-phenylalanine hydrochloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.