Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta-Methyl-gamma-octanolactone (mixture of isomers) 98.0+%, TCI America™
SDP

Catalog No. M251625ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M251625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M251625ML Supplier TCI America Supplier No. M251625ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 39212-23-2
Molecular Formula C9H16O2
Molecular Weight (g/mol) 156.23
MDL Number MFCD00209540
InChI Key WNVCMFHPRIBNCW-UHFFFAOYNA-N
Synonym 5-Butyl-4-methyldihydro-2(3H)-furanone
PubChem CID 62900
IUPAC Name 5-butyl-4-methyloxolan-2-one
SMILES CCCCC1OC(=O)CC1C

Specifications

Color Colorless
Boiling Point 64°C
Quantity 25 mL
Formula Weight 156.23
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material beta-Methyl-gamma-octanolactone (mixture of isomers)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.