Learn More
CPC Scientific H-Ala-Glu-Lys-Lys-Asp-Glu-Gly-Pro-Tyr-Arg-Met-Glu-His-Phe-Arg-Trp-Gly-Ser-Pro-Pro-Lys-Asp-OH (acetate salt) 0.5MG
SDP
Supplier:  CPC Scientific MSHS006A
Encompass
ONE-LETTER SEQUENCE: AEKKDEGPYRMEHFRWGSPPKDMOLECULAR FORMULA: C118H174N34O35S1MOLECULAR WEIGHT:2661STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [17908-57-5], SYNONYMS: melanotropinsRESEARCH AREA: Hormonal
Catalog No. 50-210-9843