Learn More
New England Biolabs, Inc. β-N-Acetylhexosaminidase f - 500 units
SDP
Catalog No. 50811836
Encompass
b-N-Acetylhexosaminidasef is an exoglycosidase that catalyzes the hydrolysis of terminal, non-reducing b-N-Acetylgalactosamine and glucosamine residues from oligosaccharides.
Catalog No. 50-811-836 Supplier New England Biolabs, Inc. Supplier No. P0721S
b-N-Acetylhexosaminidasef is an exoglycosidase that catalyzes the hydrolysis of terminal, non-reducing b-N-Acetylgalactosamine and glucosamine residues from oligosaccharides.