Learn More
New England Biolabs, Inc. β-N-Acetylglucosaminidase S - 100 units
SDP
Catalog No. 501047514
Encompass
b-N-Acetylglucosaminidase S is an exoglycosidase that catalyzes the hydrolysis of terminal, non-reducing b-N-Acetylglucosamine residues from oligosaccharides.
Catalog No. 50-104-7514 Supplier New England Biolabs, Inc. Supplier No. P0744S
b-N-Acetylglucosaminidase S is an exoglycosidase that catalyzes the hydrolysis of terminal, non-reducing b-N-Acetylglucosamine residues from oligosaccharides.