Learn More
AdipoGen beta-NAD hydrate
SDP
Catalog No. 502560391
Encompass_Preferred
Chemical. CAS 53-84-9. Formula C21H27N7O14P2 . xH2O. MW 663.43 anhydrous basis. NAD, known more formally as nicotinamide adenine dinucleotide, is a signaling molecule as well as a cofactor or substrate for many enzymes. It acts as an oxidizing agent, accepting electrons from other molecules while being converted to its reduced form, NADH. NAD is also essential for the activity of several enzymes, including polyADP-ribose polymerases and cADP-ribose synthases. For example, it is used by some sirtuins to mediate protein deacetylation, producing O-acetyl-ADP-ribose and nicotinamide as well as the deacetylated protein. NAD is an important player and factor to research metabolic pathways and pathologies.
Catalog No. 50-256-0391 Supplier AdipoGen Supplier No. CDXN0222G005
Chemical. CAS 53-84-9. Formula C21H27N7O14P2 . xH2O. MW 663.43 anhydrous basis. NAD, known more formally as nicotinamide adenine dinucleotide, is a signaling molecule as well as a cofactor or substrate for many enzymes. It acts as an oxidizing agent, accepting electrons from other molecules while being converted to its reduced form, NADH. NAD is also essential for the activity of several enzymes, including polyADP-ribose polymerases and cADP-ribose synthases. For example, it is used by some sirtuins to mediate protein deacetylation, producing O-acetyl-ADP-ribose and nicotinamide as well as the deacetylated protein. NAD is an important player and factor to research metabolic pathways and pathologies.