Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta-Naphthol Violet, TCI America™
SDP

Catalog No. N00371G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N00371G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N00371G Supplier TCI America Supplier No. N00371G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7143-21-7
Molecular Formula C16H9N3Na2O9S2
Molecular Weight (g/mol) 497.36
MDL Number MFCD00021457
InChI Key QMNPQXAVQXMXTD-UHFFFAOYSA-L
Synonym beta-naphthol violet, beta-naphtholviolet, disodium 3-hydroxy-4-e-2-4-nitrophenyl diazen-1-yl naphthalene-2,7-disulfonate, disodium 3-hydroxy-4-2-4-nitrophenyl diazen-1-yl naphthalene-2,7-disulfonate
PubChem CID 44135658
IUPAC Name disodium;4-[(4-nitrophenyl)hydrazinylidene]-3-oxonaphthalene-2,7-disulfonate
SMILES C1=CC(=CC=C1NN=C2C3=C(C=C(C=C3)S(=O)(=O)[O-])C=C(C2=O)S(=O)(=O)[O-])[N+](=O)[O-].[Na+].[Na+]

Specifications

Quantity 1 g
Formula Weight 497.36
Physical Form Crystalline Powder
Chemical Name or Material beta-Naphthol Violet
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.