Learn More
Sigma Aldrich Fine Chemicals Biosciences beta Ncotnamde mononucl250MG
Catalog No. 502907625
Encompass
beta-Nicotinamide mononucleotide (beta-NMN) is an intermediate in the nicotinamide phosphoribosyltransferase (NAMPT)-catalyzed biosynthesis of nicotinamide adenine dinucleotide (NAD). NAMPT mediates the condensation of nicotinamide with 5-phosphoribosyl-1-pyrophosphate to produce beta-NMN. beta-NMN adenyltransferase subsequently converts beta-NMN to NAD.
Catalog No. 50-290-7625 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. N3501250MG
beta-Nicotinamide mononucleotide (beta-NMN) is an intermediate in the nicotinamide phosphoribosyltransferase (NAMPT)-catalyzed biosynthesis of nicotinamide adenine dinucleotide (NAD). NAMPT mediates the condensation of nicotinamide with 5-phosphoribosyl-1-pyrophosphate to produce beta-NMN. beta-NMN adenyltransferase subsequently converts beta-NMN to NAD.