Learn More
TARGETMOL CHEMICALS INC BETA-NF-JQ1 5MG
SDP
Catalog No. 502581714
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. (beta)-NF-JQ1 is a PROTAC that recruits aryl hydrocarbon receptor E3 (AhR E3) ligase to target proteins. (beta)-NF-JQ1 uses (beta)-NF as an AhR ligand to target bromodomain (BRD)-containing proteins and induce AhR and BRD protein interactions. (beta)-NF-JQ1 shows potent anticancer activity associated with protein knockdown activity. . purity: 99%
Catalog No. 50-258-1714 Supplier TARGETMOL CHEMICALS INC Supplier No. T105265MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. (beta)-NF-JQ1 is a PROTAC that recruits aryl hydrocarbon receptor E3 (AhR E3) ligase to target proteins. (beta)-NF-JQ1 uses (beta)-NF as an AhR ligand to target bromodomain (BRD)-containing proteins and induce AhR and BRD protein interactions. (beta)-NF-JQ1 shows potent anticancer activity associated with protein knockdown activity. . purity: 99%