Learn More
Medchemexpress LLC Beta-NGF Human CHO 50ug
SDP
Catalog No. 5000179082
Encompass_Preferred
Nerve Growth Factor- (Beta-NGF NGF) is a neurotrophic factor that regulates neuronal survival and differentiation NGF is also a seminal plasma protein involved in ovulation and luteinizing NGF has potential functions in the female reproductive system to help overcome the current problem of early embryo loss[1][2][3] Beta-NGF Protein Human (CHO) is produced by CHO cells (S122-R239) with tag free
Catalog No. 50-001-79082 Supplier Medchemexpress LLC Supplier No. HYP7331650UG
Nerve Growth Factor- (Beta-NGF NGF) is a neurotrophic factor that regulates neuronal survival and differentiation NGF is also a seminal plasma protein involved in ovulation and luteinizing NGF has potential functions in the female reproductive system to help overcome the current problem of early embryo loss[1][2][3] Beta-NGF Protein Human (CHO) is produced by CHO cells (S122-R239) with tag free