Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Beta-NGF Mouse 110 100ug
SDP

Catalog No. 5000184835
Encompass_Preferred

Recombinant mouse β-NGF (110 amino acids) is a biologically active nerve growth factor provided as a lyophilized, tag-free protein for research use. It promotes neuronal survival and differentiation in cell-based assays and biochemical studies, with characterized bioactivity and recommended storage to preserve potency.

  • Recombinant mouse β-NGF, 110 amino acids.
  • Tag free.
  • Expressed in E. coli.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Lyophilized from 20 mM PB, 200 mM NaCl, pH 8.0.
  • Predicted molecular mass ~12.4 kDa.
  • Activity ED50 ≤192.19 ng/mL in TF-1 cell proliferation assay.
  • Store at -20°C; after reconstitution stable 1 week at 4°C.

Catalog No. 50-001-84835 Supplier Medchemexpress LLC Supplier No. HYP70530A100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant mouse β-NGF (110 amino acids) is a biologically active nerve growth factor provided as a lyophilized, tag-free protein for research use. It promotes neuronal survival and differentiation in cell-based assays and biochemical studies, with characterized bioactivity and recommended storage to preserve potency.

  • Recombinant mouse β-NGF, 110 amino acids.
  • Tag free.
  • Expressed in E. coli.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Lyophilized from 20 mM PB, 200 mM NaCl, pH 8.0.
  • Predicted molecular mass ~12.4 kDa.
  • Activity ED50 ≤192.19 ng/mL in TF-1 cell proliferation assay.
  • Store at -20°C; after reconstitution stable 1 week at 4°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.