Learn More
Medchemexpress LLC Beta-NGF Mouse 110 2ug
SDP
Catalog No. 5000184156
Encompass_Preferred
Recombinant mouse beta nerve growth factor (β-NGF) is provided as a 110-amino-acid His-tagged protein corresponding to residues M130-R239. Produced in Escherichia coli and purified to >95% by reducing SDS-PAGE, the protein is supplied in small vial sizes for in-lab research, with storage and handling guidance to preserve activity. Intended for research use only.
Catalog No. 50-001-84156 Supplier Medchemexpress LLC Supplier No. HYP70530A2UG
Recombinant mouse beta nerve growth factor (β-NGF) is provided as a 110-amino-acid His-tagged protein corresponding to residues M130-R239. Produced in Escherichia coli and purified to >95% by reducing SDS-PAGE, the protein is supplied in small vial sizes for in-lab research, with storage and handling guidance to preserve activity. Intended for research use only.