Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Beta-NGF protein, human (HEK293) | 98% | 2 UG
SDP

Catalog No. 5000290419
Encompass_Preferred

Recombinant human beta-nerve growth factor (beta-NGF) produced in HEK293 cells and supplied as a tag-free recombinant protein for research use. It is intended for in vitro studies of neuronal survival, differentiation, and NGF/TrkA signaling; consult the manufacturer datasheet for storage and handling instructions.

  • Produced in HEK293 cells (mammalian expression).
  • Tag-free recombinant protein.
  • Purity approximately 98% by SDS-PAGE.
  • Approximate molecular weight ~14 kDa.
  • Suitable for neuronal survival, differentiation, and signaling assays.
  • Typical quantity: 2 μg aliquot.

Catalog No. 50-002-90419 Supplier Medchemexpress LLC Supplier No. HYP76602UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human beta-nerve growth factor (beta-NGF) produced in HEK293 cells and supplied as a tag-free recombinant protein for research use. It is intended for in vitro studies of neuronal survival, differentiation, and NGF/TrkA signaling; consult the manufacturer datasheet for storage and handling instructions.

  • Produced in HEK293 cells (mammalian expression).
  • Tag-free recombinant protein.
  • Purity approximately 98% by SDS-PAGE.
  • Approximate molecular weight ~14 kDa.
  • Suitable for neuronal survival, differentiation, and signaling assays.
  • Typical quantity: 2 μg aliquot.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.