Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Beta-nerve growth factor | >90.0% | 100UG
SDP

Catalog No. 5000175926
Encompass_Preferred

Recombinant rat beta-nerve growth factor (beta-NGF) expressed in HEK293 cells, supplied as a lyophilized powder with a C-terminal His tag. The protein is formulated in PBS with 6% trehalose, characterized for purity and biological activity, and intended for research use in cell culture and in vitro assays.

  • Expressed in HEK293 cells and supplied as a lyophilized powder
  • C-terminal His tag for detection and purification
  • Formulated in PBS with 6% trehalose for stability
  • Purity >90% as determined by reducing SDS-PAGE
  • Endotoxin <1 EU/μg by LAL assay
  • Biological activity ED50 ≈1.463 ng/mL in a cell proliferation assay
  • Store at -20°C; aliquot and freeze for long-term storage

Catalog No. 50-001-75926 Supplier Medchemexpress LLC Supplier No. HYP702778100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant rat beta-nerve growth factor (beta-NGF) expressed in HEK293 cells, supplied as a lyophilized powder with a C-terminal His tag. The protein is formulated in PBS with 6% trehalose, characterized for purity and biological activity, and intended for research use in cell culture and in vitro assays.

  • Expressed in HEK293 cells and supplied as a lyophilized powder
  • C-terminal His tag for detection and purification
  • Formulated in PBS with 6% trehalose for stability
  • Purity >90% as determined by reducing SDS-PAGE
  • Endotoxin <1 EU/μg by LAL assay
  • Biological activity ED50 ≈1.463 ng/mL in a cell proliferation assay
  • Store at -20°C; aliquot and freeze for long-term storage

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.