Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Nerve growth factor-β (beta-NGF) | 86923-98-0 | MFCD00164982 | >90.0% | 20UG
SDP

Catalog No. 5000175763
Encompass_Preferred

Recombinant rat beta-nerve growth factor (beta-NGF) expressed in HEK293 cells and purified with a C-terminal His tag. Supplied lyophilized in PBS with 6% trehalose, this biologically active protein supports neuronal survival and differentiation and is suitable for in vitro cell culture assays.

  • Expressed in HEK293 cells.
  • C-terminal His tag for purification.
  • Lyophilized from PBS with 6% trehalose, pH 7.4.
  • Predicted molecular mass ~26.4 kDa.
  • Purity greater than 90% by reducing SDS-PAGE.
  • Endotoxin level <1 EU/μg as determined by LAL.
  • Stable at -20°C for long-term storage; aliquot and freeze for extended use.
  • Activity demonstrated in a TF-1 cell proliferation assay.
  • Available in multiple microgram quantities for research use.

Catalog No. 50-001-75763 Supplier Medchemexpress LLC Supplier No. HYP70277820UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant rat beta-nerve growth factor (beta-NGF) expressed in HEK293 cells and purified with a C-terminal His tag. Supplied lyophilized in PBS with 6% trehalose, this biologically active protein supports neuronal survival and differentiation and is suitable for in vitro cell culture assays.

  • Expressed in HEK293 cells.
  • C-terminal His tag for purification.
  • Lyophilized from PBS with 6% trehalose, pH 7.4.
  • Predicted molecular mass ~26.4 kDa.
  • Purity greater than 90% by reducing SDS-PAGE.
  • Endotoxin level <1 EU/μg as determined by LAL.
  • Stable at -20°C for long-term storage; aliquot and freeze for extended use.
  • Activity demonstrated in a TF-1 cell proliferation assay.
  • Available in multiple microgram quantities for research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.