Alfa Aesar™ beta-Nicotinamide adenine dinucleotide phosphate monosodium salt

Manufacturer: Alfa Aesar™ 44126MD

Quantity 250mg

 View more versions of this product

Catalog No. AA44126MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1184-16-3
    Molecular Formula C21H27N7NaO17P3
    Formula Weight 765.4
    MDL Number MFCD00036973
    Beilstein 4779954
    Synonym beta-NADP monosodium salt; Triphosphopyridine nucleotide monosodium salt
    Chemical Name or Material beta-Nicotinamide adenine dinucleotide phosphate monosodium salt

    Specifications

    Formula Weight 765.4
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 1 Warning
    Physical Form Powder
    CAS 1184-16-3
    Quantity 250mg
    Recommended Storage −30°C to −10°C
    Molecular Formula C21H27N7NaO17P3
    MDL Number MFCD00036973
    Beilstein 4779954
    Synonym beta-NADP monosodium salt; Triphosphopyridine nucleotide monosodium salt
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Color White
    Chemical Name or Material beta-Nicotinamide adenine dinucleotide phosphate monosodium salt

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    −30°C to −10°C

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit